Methyl-6-quinolyl ether ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 6-Methoxyquinoline
M0891-25G M0891-5G
TCIAM0891-25GEA 3075 SEK
TCIAM0891-25G TCIAM0891-5G
Methyl-6-quinolyl ether ≥97.0% (by GC, titration analysis)
Methyl-6-quinolyl ether
Formel: C₁₀H₉NO
MW: 159,19 g/mol
Kokpunkt: 284°C
Smältpunkt: 19 °C
Densitet: 1,152 g/cm³ (20 °C)
Storage Temperature: Kylskåp
MDL Number: MFCD00006800
CAS nummer: 5263-87-6
EINECS: 226-077-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b0f2071d9ac9d56cafee43b90b2f545cbbed8137&vid=b0f2071d9ac9d56cafee43b90b2f545cbbed8137&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %