8-Aminoquinoline ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Quinolin-8-ylamine
A0419-25G
A0419-5G
A0419-100G
TCIAA0419-25GEA
1275
SEK
TCIAA0419-25G
TCIAA0419-5G
TCIAA0419-100G
8-Aminoquinoline ≥98.0% (by GC, titration analysis)
8-Aminoquinoline
Formel:
C₉H₈N₂ MW: 144,18 g/mol Kokpunkt: 145°C Smältpunkt: 64 °C Densitet: 1,337 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00006809 CAS nummer: 578-66-5 EINECS: 209-427-9 |
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 63.0 to 67.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...