6-Aminoquinoline ≥99.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Quinolin-6-ylamine

A1370-1G A1370-5G
TCIAA1370-1GEA 365 SEK
TCIAA1370-1G TCIAA1370-5G
6-Aminoquinoline ≥99.0%
6-Aminoquinoline
Formel: C₉H₈N₂
MW: 144,18 g/mol
Kokpunkt: 187°C
Smältpunkt: 116…117 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006803
CAS nummer: 580-15-4
EINECS: 209-453-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09791732cde54b0ed2d7e0aad11dc638efc943cb&vid=f9798de392e7962a0cc78dd4d015d0491d329da6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 114.0 to 118.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR