5-Aminoquinoline ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Quinolin-5-amine, Quinolin-5-ylamine
A0959-5G
A0959-25G
TCIAA0959-5GEA
535
SEK
TCIAA0959-5G
TCIAA0959-25G
5-Aminoquinoline ≥99.0%
5-Aminoquinoline
Formel:
C₉H₈N₂ MW: 144,18 g/mol Kokpunkt: 310°C Smältpunkt: 107…109 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00006797 CAS nummer: 611-34-7 EINECS: 210-266-1 |
Specifikation testresultat
Appearance | White to Yellow to Orange powder to crystalline |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 108.0 to 112.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...