6-Bromoquinoline ≥95.0%
Leverantör: TCI
|
Varning
|
B2015-1G
B2015-5G
TCIAB2015-1GEA
365
SEK
TCIAB2015-1G
TCIAB2015-5G
6-Bromoquinoline ≥95.0%
6-Bromoquinoline
Formel:
C₉H₆BrN Kokpunkt: 116°C Storage Temperature: Kylskåp |
MDL Number:
MFCD00024023 CAS nummer: 5332-25-2 EINECS: 226-238-7 |
|
Specifikation testresultat
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...