(1R)-(+)-α-Pinene ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (+)-α-Pinene, D(+)-α-Pinene, (1R)-(+)-2-Pinene, (+)-Pin-2(3)-ene
P1099-25ML P1099-500ML
TCIAP1099-25MLEA 230 SEK
TCIAP1099-25ML TCIAP1099-500ML
(1R)-(+)-α-Pinene ≥97.0%
(1R)-(+)-α-Pinene
Formel: C₁₀H₁₆
MW: 136,24 g/mol
Kokpunkt: 156°C
Smältpunkt: –62 °C
Densitet: 0,857 g/cm³ (20 °C)
Flampunkt: 32°C
Storage Temperature: Kylskåp
MDL Number: MFCD00001346
CAS nummer: 7785-70-8
EINECS: 232-087-8
UN: 2368
ADR: 3,III
Merck Index: 14,07445
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9af43c20dc6b34bcb08a3b9ecc4b6138cd7384d6&vid=ffae0253358eeb1447a79fc7fd6f7be32323ac25&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %
Specific rotation [a]20/D +42.0 to +46.0 deg-C (neat)