4-Phenylbenzoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: Biphenyl-4-carboxylic acid, [1,1'-Biphenyl]-4-carboxylic acid, 4-Biphenylcarboxylic acid
P0961-25G P0961-100G
TCIAP0961-25GEA 325 SEK
TCIAP0961-25G TCIAP0961-100G
4-Phenylbenzoic acid ≥98.0% (by GC, titration analysis)
4-Phenylbenzoic acid
Formel: C₁₃H₁₀O₂
MW: 198,22 g/mol
Kokpunkt: 373 °C (1013 hPa)
Smältpunkt: 220 °C
Densitet: 1,284 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002553
CAS nummer: 92-92-2
EINECS: 202-203-1
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 225.0 to 229.0 deg-C
Solubility in hot Methanol very faint turbidity