4-Phenylcyclohexanone ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 4-Phenylcyclohexan-1-one, 1-Oxo-4-phenylcyclohexane
P0975-25G P0975-250G
TCIAP0975-25GEA 750 SEK
TCIAP0975-25G TCIAP0975-250G
4-Phenylcyclohexanone ≥98.0%
4-Phenylcyclohexanone
Formel: C₁₂H₁₄O
MW: 174,24 g/mol
Kokpunkt: 294 °C (1013 hPa)
Smältpunkt: 76…78 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001641
CAS nummer: 4894-75-1
EINECS: 225-517-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ffc30a8cde9c6bbd2cb8c782a82ae25121da879f&vid=86982824c0139b51fa6b73848aaedcb63065687d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 80.0 deg-C
Solubility in Methanol almost transparency