1,2,4,5-Tetrametylbensen ≥98.0%
Leverantör: TCI EUROPE
T0714-25G T0714-500G
TCIAT0714-25GEA 255 SEK
TCIAT0714-25G TCIAT0714-500G
1,2,4,5-Tetrametylbensen ≥98.0%
1,2,4,5-Tetrametylbensen
Formel: C₁₀H₁₄
MW: 134,22 g/mol
Kokpunkt: 197°C
Smältpunkt: 78…81 °C
Densitet: 1,036 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008528
CAS nummer: 95-93-2
EINECS: 202-465-7
ADR: 4.1,II
Merck Index: 14,03468
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4ac9fad5abf10a1513055620e8be1a60dbf6ef1&vid=94aa500e015fe8a88530aaded54bb936df2f007b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 81.0 deg-C
Solubility in hot Methanol almost transparency