1-Iodo-2,3,5,6-tetramethylbenzene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 2,3,5,6-Tetramethyliodobenzene, 3-Iodo-1,2,4,5-tetramethylbenzene

T0992-5G T0992-1G
TCIAT0992-5GEA 1275 SEK
TCIAT0992-5G TCIAT0992-1G
1-Iodo-2,3,5,6-tetramethylbenzene ≥98.0%
1-Iodo-2,3,5,6-tetramethylbenzene
Formel: C₁₀H₁₃I
MW: 260,12 g/mol
Kokpunkt: 290°C
MDL Number: MFCD00013705
CAS nummer: 2100-25-6
EINECS: 218-266-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62609acd926b107dbb32e7bc9c4b9f54b2839f31&vid=62609acd926b107dbb32e7bc9c4b9f54b2839f31&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 82.0 deg-C
Solubility in hot Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR