2,4,6-Trimethylphenol ≥98.0%

Leverantör: TCI EUROPE
Fara

T0486-25G T0486-500G T0486-100G
TCIAT0486-25GEA 1475 SEK
TCIAT0486-25G TCIAT0486-500G TCIAT0486-100G
2,4,6-Trimethylphenol ≥98.0%
2,4,6-Trimethylphenol
Formel: C₉H₁₂O
MW: 136,19 g/mol
Kokpunkt: 95°C
Smältpunkt: 74 °C
Densitet: 1,046 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002235
CAS nummer: 527-60-6
EINECS: 208-419-2
UN: 2430
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b672ed2ea13bc3ef36db3368e5bcd00a97403d4&vid=c5efc8f1beb88364c78bce9f941a5c0a671064a7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR