2,3,6-Trimethylphenol ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Hydroxypseudocumene
T0877-25G
T0877-500G
TCIAT0877-25GEA
230
SEK
TCIAT0877-25G
TCIAT0877-500G
2,3,6-Trimethylphenol ≥98.0%
2,3,6-Trimethylphenol
Formel:
C₉H₁₂O MW: 136,19 g/mol Kokpunkt: 120°C Smältpunkt: 63…64 °C Densitet: 1,189 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00002229 CAS nummer: 2416-94-6 EINECS: 219-330-3 UN: 1759 ADR: 8,III |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 62.0 to 66.0 deg-C |
Solubility in Methanol | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...