2,4,6-Tri-tert-butylaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
2,4,6-Tris(tert-butyl)aniline
T1694-1G
T1694-5G
TCIAT1694-1GEA
510
SEK
TCIAT1694-1G
TCIAT1694-5G
2,4,6-Tri-tert-butylaniline ≥98.0% (by GC, titration analysis)
2,4,6-Tri-tert-butylaniline
Formel:
C₁₈H₃₁N MW: 261.45 g/mol Kokpunkt: 302 Smältpunkt: 145 |
MDL Number:
MFCD00011645 CAS nummer: 961-38-6 EINECS: 213-507-9 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 145.0 to 149.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...