1,4-Diethylbenzene ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
D0479-25ML
D0479-5ML
TCIAD0479-25MLEA
1150
SEK
TCIAD0479-25ML
TCIAD0479-5ML
1,4-Diethylbenzene ≥98.0%
1,4-Diethylbenzene
Formel:
C₁₀H₁₄ MW: 134,22 g/mol Kokpunkt: 184°C Smältpunkt: –43°C Densitet: 0,862 g/cm³ (20 °C) Flampunkt: 55°C Storage Temperature: Omgivande |
MDL Number:
MFCD00009264 CAS nummer: 105-05-5 EINECS: 203-265-2 UN: 2049 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...