1,4-Di-tert-butylbenzene ≥99.0%
Leverantör: TCI EUROPE
Varning
D2111-25G D2111-5G
TCIAD2111-25GEA 1750 SEK
TCIAD2111-25G TCIAD2111-5G
1,4-Di-tert-butylbenzene ≥99.0%
1,4-Di-tert-butylbenzene
Formel: C₁₄H₂₂
MW: 190,33 g/mol
Kokpunkt: 236°C
Smältpunkt: 76…79 °C
Densitet: 0,95 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00008836
CAS nummer: 1012-72-2
EINECS: 213-790-9
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b932d801d7b4f019be55b221b985ad7ac3cb3491&vid=b932d801d7b4f019be55b221b985ad7ac3cb3491&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 77.0 to 80.0 deg-C
Solubility in hot Methanol almost transparency