1,3-Diisopropylbenzene ≥85.0%

Leverantör: TCI
Varning

D0927-25ML D0927-500ML
TCIAD0927-25MLEA 215 SEK
TCIAD0927-25ML TCIAD0927-500ML
1,3-Diisopropylbenzene ≥85.0%
1,3-Diisopropylbenzene
Formel: C₁₂H₁₈
Kokpunkt: 203°C
Smältpunkt: –40 °C
Densitet: 0,85593 g/cm³ (20 °C)
Flampunkt: 77°C
Storage Temperature: Omgivande
MDL Number: MFCD00008889
CAS nummer: 99-62-7
EINECS: 202-773-1
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab73b113a0b0f0102f2ae7c8ccf966836de571db&vid=753d730aab332e79dd0563bc03d01b55257432b6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR