o-Xylen ≥98.0%
Leverantör: TCI
|
Varning
|
X0012-25ML
X0012-500ML
TCIAX0012-25MLEA
230
SEK
TCIAX0012-25ML
TCIAX0012-500ML
o-Xylen ≥98.0%
o-Xylen
Formel:
C₈H₁₀ Kokpunkt: 145°C Smältpunkt: –25 °C Densitet: 0,88 g/cm³ (20 °C) Flampunkt: 33°C (closed cup) Storage Temperature: Omgivande |
MDL Number:
MFCD00008519 CAS nummer: 95-47-6 EINECS: 202-422-2 UN: 1307 ADR: 3,III Merck Index: 14,10081 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...