3,5-Xylenol ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 3,5-Dimetylfenol
D1829-25G D1829-100G
TCIAD1829-25GEA 335 SEK
TCIAD1829-25G TCIAD1829-100G
3,5-Xylenol ≥98.0%
3,5-Xylenol
Formel: C₈H₁₀O
MW: 122,17 g/mol
Kokpunkt: 95°C
Smältpunkt: 61…64 °C
Densitet: 0,968 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002307
CAS nummer: 108-68-9
EINECS: 203-606-5
UN: 2261
ADR: 6.1,II
Merck Index: 14,10082
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a428f9014c437e5801397e5d739728c0951410c&vid=1852fcab2fac30bc8ec6243885af6bfb405663e4&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 66.0 deg-C