2,4-Di-tert-butylphenol ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 1,3-Di(dimethylethyl)-4-hydroxybenzene

D0229-25G D0229-500G
TCIAD0229-25GEA 215 SEK
TCIAD0229-25G TCIAD0229-500G
2,4-Di-tert-butylphenol ≥97.0%
2,4-Di-tert-butylphenol
Formel: C₁₄H₂₂O
MW: 206,33 g/mol
Kokpunkt: 265 °C (1013 hPa)
Smältpunkt: 53…56 °C
Densitet: 0,887 g/cm³ (20 °C)
Flampunkt: 129 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008828
CAS nummer: 96-76-4
EINECS: 202-532-0
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=693ccb1f321740f894e4e2eb66ee29e4b4b7c86e&vid=a99d02b5d7bcf642572980cfb01001a55c89c85a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 56.0 to 59.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR