3,5-Xylidin ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Fara
Synonyms: 3,5-Dimethylaniline, 5-Amino-m-xylene
D0671-25ML D0671-500ML
TCIAD0671-25MLEA 215 SEK
TCIAD0671-25ML TCIAD0671-500ML
3,5-Xylidin ≥98.0% (by GC, titration analysis)
3,5-Xylidin
Formel: C₈H₁₁N
MW: 121.18 g/mol
Kokpunkt: 221°C
Smältpunkt: 9.8
Flampunkt: 97°C
Storage Temperature: Omgivande
MDL Number: MFCD00007813
CAS nummer: 108-69-0
EINECS: 203-607-0
UN: 1711
ADR: 6.1 II
Merck Index: 14,10084
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23ae08a33605c4d5149064442955d16a88f7b860&vid=f106ab6d6888c595cc88d3d92b1eff469f688fb5&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %