2,5-Xylidin ≥97.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
2-Amino-p-xylene, 2,5-Dimethylaniline
D0668-25G
D0668-500G
TCIAD0668-25GEA
215
SEK
TCIAD0668-25G
TCIAD0668-500G
2,5-Xylidin ≥97.0% (by GC, titration analysis)
2,5-Xylidin
Formel:
C₈H₁₁N MW: 121,18 g/mol Kokpunkt: 104°C Smältpunkt: 15 °C Densitet: 0,979 g/cm³ (20 °C) Flampunkt: 106°C Storage Temperature: Omgivande |
MDL Number:
MFCD00007743 CAS nummer: 95-78-3 EINECS: 202-451-0 UN: 1711 ADR: 6.1,II Merck Index: 14,10084 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...