2,4-Xylidin ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Fara
|
Synonyms:
2,4-Dimethylaniline, 4-Amino-m-xylene, m-Xylidine, 2,4-Xylidine
D0667-25ML
D0667-500ML
TCIAD0667-25MLEA
215
SEK
TCIAD0667-25ML
TCIAD0667-500ML
2,4-Xylidin ≥98.0% (by GC, titration analysis)
2,4-Xylidin
Formel:
C₈H₁₁N Kokpunkt: 218 °C (1013 hPa) Smältpunkt: –16 °C Densitet: 0,98 g/cm³ (20 °C) Flampunkt: 90°C Storage Temperature: Omgivande |
MDL Number:
MFCD00007738 CAS nummer: 95-68-1 EINECS: 202-440-0 UN: 1711 ADR: 6.1,II Merck Index: 14,10084 |
|
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...