4-tert-Butylbenzylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: p-tert-Butylbenzylamine
B1587-25ML B1587-5ML
TCIAB1587-25MLEA 2000 SEK
TCIAB1587-25ML TCIAB1587-5ML
4-tert-Butylbenzylamine ≥98.0% (by GC, titration analysis)
4-tert-Butylbenzylamine
Formel: C₁₁H₁₇N
MW: 163,26 g/mol
Kokpunkt: 124°C
Flampunkt: 107°C
MDL Number: MFCD00040754
CAS nummer: 39895-55-1
EINECS: 254-681-6
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7a000513e924c663d2be5e53da5319374bc5644&vid=4df653d2b0c895b9d12e9198cdb8d0b61222e13c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %