α,α,α,α',α',α'-Hexafluoro-m-xylene ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 1,3-Bis(trifluoromethyl)benzene, α,α,α,β,β,β-Hexafluoro-m-xylene
H0585-25G H0585-500G H0585-100G
TCIAH0585-25GEA 215 SEK
TCIAH0585-25G TCIAH0585-500G TCIAH0585-100G
α,α,α,α',α',α'-Hexafluoro-m-xylene ≥98.0%
α,α,α,α',α',α'-Hexafluoro-m-xylene
Formel: C₈H₄F₆
MW: 214,11 g/mol
Kokpunkt: 116°C
Smältpunkt: –35 °C
Densitet: 1,383 g/cm³ (22 °C)
Flampunkt: 26°C
Storage Temperature: Omgivande
MDL Number: MFCD00000392
CAS nummer: 402-31-3
EINECS: 206-939-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e19c77f23b3f581b2366df7d6a4b7116968fb284&vid=b87397f93be6a056cb6e7765e50dff5367964d82&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %