4-tert-Butyl-α-chlorotoluene ≥89.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
p-tert-Butylbenzyl chloride, 4-tert-Butylbenzyl chloride, p-tert-Butyl-α-chlorotoluene, 1-(tert-Butyl)-4-(chloromethyl)benzene
B2235-25G
B2235-5G
TCIAB2235-25GEA
1400
SEK
TCIAB2235-25G
TCIAB2235-5G
4-tert-Butyl-α-chlorotoluene ≥89.0%
4-tert-Butyl-α-chlorotoluene
Formel:
C₁₁H₁₅Cl MW: 182,69 g/mol Flampunkt: 94°C |
MDL Number:
MFCD00000918 CAS nummer: 19692-45-6 EINECS: 243-228-8 UN: 1760 ADR: 8,III |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 89.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...