α-Chloro-m-xylene ≥97.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1-(Chloromethyl)-3-methylbenzene, 3-Methylbenzyl chloride

C0865-25G C0865-500G
TCIAC0865-25GEA 295 SEK
TCIAC0865-25G TCIAC0865-500G
α-Chloro-m-xylene ≥97.0%
α-Chloro-m-xylene
Formel: C₈H₉Cl
MW: 140,61 g/mol
Kokpunkt: 196°C
Densitet: 1,064 g/cm³ (20 °C)
Flampunkt: 76°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000909
CAS nummer: 620-19-9
EINECS: 210-628-9
UN: 3265
ADR: 8,III
Merck Index: 14,10091
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9276e8694987bb5900adff1d9fd643d74adb5095&vid=ac97b3eb6da1b71e63c417804e96b13595d354c5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR