α-Chloro-o-xylene ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2-Methylbenzyl chloride
C0864-25G C0864-500G
TCIAC0864-25GEA 215 SEK
TCIAC0864-25G TCIAC0864-500G
α-Chloro-o-xylene ≥98.0%
α-Chloro-o-xylene
Formel: C₈H₉Cl
MW: 140,61 g/mol
Kokpunkt: 198°C
Smältpunkt: –2 °C
Densitet: 1,08 g/cm³ (20 °C)
Flampunkt: 97°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000899
CAS nummer: 552-45-4
EINECS: 209-013-8
UN: 3265
ADR: 8,III
Merck Index: 14,10091
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dfe368a0d8d35319390a47b6b0a6ce80b909158a&vid=2969f3a34b0888435a097547f73cf45aea056e6d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %