2,5-Dimethylbenzoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
D1433-10G D1433-25G
TCIAD1433-10GEA 590 SEK
TCIAD1433-10G TCIAD1433-25G
2,5-Dimethylbenzoic acid ≥98.0% (by GC, titration analysis)
2,5-Dimethylbenzoic acid
Formel: C₉H₁₀O₂
Kokpunkt: 268 °C (1013 hPa)
Smältpunkt: 131…135 °C
Densitet: 1,06 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002482
CAS nummer: 610-72-0
EINECS: 210-235-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5be1701a919f8a3633532bf1d3da49db09a97de8&vid=9d8e062dd5d197ff9e1b6265de1e8b290b08491a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 132.0 to 135.0 deg-C
Solubility in Methanol very faint turbidity