3-Methyl-2-thenoic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Methylthiophene-2-carboxylic acid

M1459-25G M1459-5G
TCIAM1459-25GEA 1350 SEK
TCIAM1459-25G TCIAM1459-5G
3-Methyl-2-thenoic acid ≥98.0%
3-Methyl-2-thenoic acid
Formel: C₆H₆O₂S
MW: 142,18 g/mol
MDL Number: MFCD00005438
CAS nummer: 23806-24-8
EINECS: 245-894-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d955ed8a54ab73068142d01494643c0a28ce071&vid=9d955ed8a54ab73068142d01494643c0a28ce071&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 143.0 to 148.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR