5-Bromothiophene-2-carbaldehyde ≥97.0%
Leverantör: TCI
Synonyms: 5-Bromo-2-thiophenecarboxaldehyde
B1135-25G B1135-5G
TCIAB1135-25GEA 1475 SEK
TCIAB1135-25G TCIAB1135-5G
5-Bromothiophene-2-carbaldehyde ≥97.0%
5-Bromothiophene-2-carbaldehyde
Formel: C₅H₃BrOS
MW: 191.05 g/mol
Kokpunkt: 115°C
Flampunkt: 98°C
Storage Temperature: Kylskåp
MDL Number: MFCD00005432
CAS nummer: 4701-17-1
EINECS: 225-176-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76050205cd53586b900a1a210d7e5f7fd85ff937&vid=76050205cd53586b900a1a210d7e5f7fd85ff937&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange powder to lump
Purity (GC) min. 97.0 %