(R,R)-(+)-Hydrobenzoin ≥99.0%
Leverantör: TCI EUROPE
Synonyms:
(1R,2R)-1,2-Diphenylethane-1,2-diol, (R,R)-(+)-1,2-Diphenyl-1,2-ethanediol
H0818-25G
H0818-5G
TCIAH0818-25GEA
3200
SEK
TCIAH0818-25G
TCIAH0818-5G
(R,R)-(+)-Hydrobenzoin ≥99.0%
(R,R)-(+)-Hydrobenzoin
Formel:
C₁₄H₁₄O₂ MW: 214,26 g/mol |
MDL Number:
MFCD00064254 CAS nummer: 52340-78-0 Merck Index: 14,04777 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 146.0 to 150.0 deg-C |
Specific rotation [a]20/D | +90.0 to +96.0 deg (C=2, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...