(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 99,0 GC, T

Leverantör: TCI
Varning

A1231-1G A1231-5G
TCIAA1231-1GEA 335 SEK
TCIAA1231-1G TCIAA1231-5G
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol 99,0 GC, T
(1S,2R)-(+)-2-Amino-1,2-diphenylethanol
Formel: C₁₄H₁₅NO
MW: 213.28 g/mol
MDL Number: MFCD00074959
CAS nummer: 23364-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1145a66b6eb0809a86bd02ae0d5c84add2a4d725&vid=1145a66b6eb0809a86bd02ae0d5c84add2a4d725&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 143.0 to 146.0 deg-C
Specific rotation [a]20/D +6.0 to +8.0 deg (C=0.6, EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR