(R)-(+)-4-Bromo-α-methylbenzylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: (R)-1-(4-Bromophenyl)ethylamine, (R)-(+)-1-(4-Bromophenyl)ethylamine

B3672-1G B3672-5G
TCIAB3672-1GEA 615 SEK
TCIAB3672-1G TCIAB3672-5G
(R)-(+)-4-Bromo-α-methylbenzylamine ≥98.0% (by GC, titration analysis)
(R)-(+)-4-Bromo-α-methylbenzylamine
Formel: C₈H₁₀BrN
MW: 200,08 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00066025
CAS nummer: 45791-36-4
EINECS: 256-245-0
UN: 2735
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f1c17b260f027e3904b341a2c5b5bdd388bfbfeb&vid=f1c17b260f027e3904b341a2c5b5bdd388bfbfeb&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR