4-Aminomorpholine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Fara
|
A0832-25ML
A0832-5ML
TCIAA0832-25MLEA
1475
SEK
TCIAA0832-25ML
TCIAA0832-5ML
4-Aminomorpholine ≥98.0% (by GC, titration analysis)
4-Aminomorpholine
Formel:
C₄H₁₀N₂O Kokpunkt: 168°C Flampunkt: 58°C |
MDL Number:
MFCD00006174 CAS nummer: 4319-49-7 EINECS: 224-347-4 UN: 2733 ADR: 3,III |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...