Dimethyl hexafluoroglutarate ≥96.0%

Leverantör: TCI

D3589-1G D3589-5G
TCIAD3589-1GEA 255 SEK
TCIAD3589-1G TCIAD3589-5G
Dimethyl hexafluoroglutarate ≥96.0%
Dimethyl hexafluoroglutarate
Formel: C₇H₆F₆O₄
Kokpunkt: 100°C
Smältpunkt: –32°C
MDL Number: MFCD00155851
CAS nummer: 1513-62-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=955c25037a084e87b90fefb80607bb1da9006110&vid=955c25037a084e87b90fefb80607bb1da9006110&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR