4-Hydroxy-3,5-dimethylbenzaldehyde ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3,5-Dimethyl-4-hydroxybenzaldehyde

H1023-25G H1023-5G
TCIAH1023-25GEA 1125 SEK
TCIAH1023-25G TCIAH1023-5G
4-Hydroxy-3,5-dimethylbenzaldehyde ≥98.0%
4-Hydroxy-3,5-dimethylbenzaldehyde
Formel: C₉H₁₀O₂
MW: 150,18 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00006946
CAS nummer: 2233-18-3
EINECS: 218-774-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49b1509fc13444aedc5bd3194e54987dc64ddf02&vid=12a2d2e70adef9c6ccc40349dce18c4d66b48b90&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR