Butyraldehydoxim ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Butyraldoxime

B0753-25ML B0753-500ML
TCIAB0753-25MLEA 965 SEK
TCIAB0753-25ML TCIAB0753-500ML
Butyraldehydoxim ≥95.0%
Butyraldehydoxim
Formel: C₄H₉NO
Flampunkt: 58°C
MDL Number: MFCD00013943
CAS nummer: 110-69-0
EINECS: 203-792-8
UN: 2840
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d27e3564fc5f9c048106888eee9dcf40ce42c6e&vid=0d27e3564fc5f9c048106888eee9dcf40ce42c6e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR