4-Bromophenethylamine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: 2-(4-Bromophenyl)ethylamine

B3454-1G B3454-5G
TCIAB3454-1GEA 350 SEK
TCIAB3454-1G TCIAB3454-5G
4-Bromophenethylamine ≥98.0% (by GC, titration analysis)
4-Bromophenethylamine
Formel: C₈H₁₀BrN
MW: 200,08 g/mol
Kokpunkt: 72°C
MDL Number: MFCD00008189
CAS nummer: 73918-56-6
EINECS: 277-636-2
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8268698ae2b75ab48ba79b51d98ead26199ef2ae&vid=8268698ae2b75ab48ba79b51d98ead26199ef2ae&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR