Benzoin ethyl ether ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Benzoin Ethyl Ether, O-Ethylbenzoin
B1019-25G
B1019-500G
B1019-100G
TCIAB1019-25GEA
325
SEK
TCIAB1019-25G
TCIAB1019-500G
TCIAB1019-100G
Benzoin ethyl ether ≥99.0%
Benzoin ethyl ether
Formel:
C₁₆H₁₆O₂ MW: 240,3 g/mol |
MDL Number:
MFCD00009242 CAS nummer: 574-09-4 EINECS: 209-366-8 UN: 3077 ADR: 9,III Merck Index: 14,01093 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 56.0 to 61.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...