Chlorodiisopropylsilane ≥95.0%

Leverantör: TCI
Fara

Synonyms: Diisopropylchlorosilane

C1492-25ML C1492-5ML
TCIAC1492-25MLEA 4350 SEK
TCIAC1492-25ML TCIAC1492-5ML
Chlorodiisopropylsilane ≥95.0%
Chlorodiisopropylsilane
Formel: C₆H₁₅ClSi
MW: 150.72 g/mol
Kokpunkt: 143°C
Flampunkt: 22°C
MDL Number: MFCD00054896
CAS nummer: 2227-29-4
UN: 2985
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce25ca5857774ea3e033a608d828032052c51133&vid=ce25ca5857774ea3e033a608d828032052c51133&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR