2,4-Dimethylbenzaldehyde ≥97.0%

Leverantör: TCI EUROPE

D2392-25G D2392-500G
TCIAD2392-25GEA 350 SEK
TCIAD2392-25G TCIAD2392-500G
2,4-Dimethylbenzaldehyde ≥97.0%
2,4-Dimethylbenzaldehyde
Formel: C₉H₁₀O
MW: 134,18 g/mol
Kokpunkt: 98°C
Smältpunkt: –9 °C
Densitet: 0,96 g/cm³ (20 °C)
Flampunkt: 89°C
Storage Temperature: Omgivande
MDL Number: MFCD00003340
CAS nummer: 15764-16-6
EINECS: 239-856-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97d0c7188bd2755ba5b12808caec76988d58198b&vid=60463b13a8d73d69f721b48b93f8141538338255&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR