α,α'-Dibromo-o-xylene ≥98.0%

Leverantör: TCI
Fara

Synonyms: 1,2-Bis(bromomethyl)benzene, o-Xylylene bromide, o-Xylylene dibromide, alpha,alpha'-Dibromo-o-xylene, o-Xylene dibromide

D0214-25G D0214-100G
TCIAD0214-25GEA 615 SEK
TCIAD0214-25G TCIAD0214-100G
α,α'-Dibromo-o-xylene ≥98.0%
α,α'-Dibromo-o-xylene
Formel: C₈H₈Br₂
Kokpunkt: 140°C
Smältpunkt: 90…93 °C
Densitet: 1,96 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00000175
CAS nummer: 91-13-4
EINECS: 202-042-7
UN: 2928
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec6ee5ea723bfefba680edea056a2ef73c65b42b&vid=ec6ee5ea723bfefba680edea056a2ef73c65b42b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 96.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR