3-Methylbiphenyl ≥95.0%
Leverantör: TCI EUROPE
Synonyms:
3-Phenyltoluene
M0296-1G
M0296-25G
M0296-5G
TCIAM0296-1GEA
390
SEK
TCIAM0296-1G
TCIAM0296-25G
TCIAM0296-5G
3-Methylbiphenyl ≥95.0%
3-Methylbiphenyl
Formel:
C₁₃H₁₂ MW: 168,24 g/mol Kokpunkt: 272°C Smältpunkt: 4,53 °C Densitet: 1,0319 g/cm³ (20 °C) |
MDL Number:
MFCD00008533 CAS nummer: 643-93-6 EINECS: 211-404-3 |
Specifikation testresultat
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...