2-Methylbiphenyl ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Phenyltoluene

M1242-1G M1242-5G
TCIAM1242-1GEA 645 SEK
TCIAM1242-1G TCIAM1242-5G
2-Methylbiphenyl ≥95.0%
2-Methylbiphenyl
Formel: C₁₃H₁₂
MW: 168,24 g/mol
Kokpunkt: 256°C
Smältpunkt: –0,2 °C
Densitet: 1,011 g/cm³ (20 °C)
Flampunkt: 107°C
MDL Number: MFCD00008517
CAS nummer: 643-58-3
EINECS: 211-400-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc7a53a88557503cdd1ec1f6c747bac8bca89e37&vid=3837bb105a5d69b45b086deb3fcdc1af7817e33b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR