Dichloro(diphenyl)methane ≥98.0%
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
Dichlorodiphenylmethane, α,α-Dichlorodiphenylmethane
D0361-25G
D0361-250G
TCIAD0361-25GEA
550
SEK
TCIAD0361-25G
TCIAD0361-250G
Dichloro(diphenyl)methane ≥98.0%
Dichloro(diphenyl)methane
Formel:
(C₆H₅)₂CCl₂ MW: 237,13 g/mol Kokpunkt: 193°C Smältpunkt: 148 °C Densitet: 1,25 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00000811 CAS nummer: 2051-90-3 EINECS: 218-134-5 UN: 1760 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...