Chloro(diphenyl)methane ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Benzhydryl chloride

B1078-25ML B1078-500ML
TCIAB1078-25MLEA 510 SEK
TCIAB1078-25ML TCIAB1078-500ML
Chloro(diphenyl)methane ≥95.0%
Chloro(diphenyl)methane
Formel: (C₆H₅)₂CHCl
MW: 202,68 g/mol
Kokpunkt: 116°C
Smältpunkt: ∼17 °C
Densitet: 1,1346 g/cm³ (20 °C)
Flampunkt: 160°C
Storage Temperature: Omgivande
MDL Number: MFCD00000855
CAS nummer: 90-99-3
EINECS: 202-031-7
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a5a928e4de96549cff4e41529ded026fb39d60fe&vid=bdeb50d1c1eab0a2b2a654bff699ba9ec970ed38&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to lump
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR