1,1-Diphenylacetone ≥99.0%

Leverantör: TCI EUROPE

Synonyms: asym. Diphenylacetone, 1,1-Diphenyl-2-propanone

D1397-10G D1397-25G
TCIAD1397-10GEA 565 SEK
TCIAD1397-10G TCIAD1397-25G
1,1-Diphenylacetone ≥99.0%
1,1-Diphenylacetone
Formel: (C₆H₅)₂CHC(O)CH₃
MW: 210,28 g/mol
Kokpunkt: 135°C
Smältpunkt: 59…62 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00008759
CAS nummer: 781-35-1
EINECS: 212-307-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c50aecbc608591bddaab6e278cb78a095b1c0dda&vid=8345ebfa056d000383fa43d864f24be7552b646d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 59.0 to 62.0 deg-C
Solubility in Methanol within almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR