Propylbenzen ≥99.0%

Leverantör: TCI
Fara

P0523-25ML P0523-500ML P0523-100ML
TCIAP0523-25MLEA 255 SEK
TCIAP0523-25ML TCIAP0523-500ML TCIAP0523-100ML
Propylbenzen ≥99.0%
Propylbenzen
Formel: C₉H₁₂
Kokpunkt: 159°C
Smältpunkt: –99 °C
Densitet: 0,862 g/cm³ (20 °C)
Flampunkt: 30°C
Storage Temperature: Omgivande
MDL Number: MFCD00009377
CAS nummer: 103-65-1
EINECS: 203-132-9
UN: 2364
ADR: 3,III
Merck Index: 14,07844
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=624ba8f0120d78673f526e2035e5a5a0cf7efe5c&vid=50191af7e7969353c2861a6dc68d976b00898ef4&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR