Pentylbenzene ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
1-Phenylpentane, Amylbenzene
A0449-25ML
A0449-500ML
TCIAA0449-25MLEA
605
SEK
TCIAA0449-25ML
TCIAA0449-500ML
Pentylbenzene ≥98.0%
Pentylbenzene
Formel:
C₁₁H₁₆ MW: 148.25 g/mol Kokpunkt: 205°C Smältpunkt: –75°C Flampunkt: 66°C Storage Temperature: Omgivande |
MDL Number:
MFCD00009502 CAS nummer: 538-68-1 EINECS: 208-701-5 UN: 3082 ADR: 9 III Merck Index: 14,00600 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...