Octylbenzene ≥98.0%

Leverantör: TCI EUROPE

Synonyms: 1-Phenyloctane

O0046-25ML O0046-500ML O0046-100ML
TCIAO0046-25MLEA 735 SEK
TCIAO0046-25ML TCIAO0046-500ML TCIAO0046-100ML
Octylbenzene ≥98.0%
Octylbenzene
Formel: C₁₄H₂₂
MW: 190,33 g/mol
Kokpunkt: 264°C
Smältpunkt: –3 °C
Densitet: 0,8562 g/cm³ (20 °C)
Flampunkt: 107°C
Storage Temperature: Omgivande
MDL Number: MFCD00009564
CAS nummer: 2189-60-8
EINECS: 218-582-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8bcafb144c86228f4b51240d279548ced56970b4&vid=b7a76249c68ec6be1a5e7e18efb1fed3e8178182&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR